Sterlisation flat pouches, Stericlin®
Lieferant: VP Medical Packaging
3FKFB210184 3FKFB210176 3FKFB210166 3FKFB210168 3FKFB210156 3FKFB210126 3FKFB210144 3FKFB210172 3FKFB210180 3FKFB210116 3FKFB210128 3FKFB210102 3FKFB210110 3FKFB210104 3FKFB210118 3FKFB210148 3FKFB210122 3FKFB210152 3FKFB210106 3FKFB210108 3FKFB210112 3FKFB210114 3FKFB210120 3FKFB210124 3FKFB210130 3FKFB210132 3FKFB210134 3FKFB210136 3FKFB210138 3FKFB210140 3FKFB210142 3FKFB210146 3FKFB210150 3FKFB210154 3FKFB210158 3FKFB210160 3FKFB210162 3FKFB210164 3FKFB210170 3FKFB210178 3FKFB210182
129-1262EA 270 EUR
129-1262 129-1258 129-1253 129-1254 129-1248 129-1233 129-1242 129-1256 129-1260 129-1228 129-1234 129-1221 129-1225 129-1222 129-1229 129-1244 129-1231 129-1246 129-1223 129-1224 129-1226 129-1227 129-1230 129-1232 129-1235 129-1236 129-1237 129-1238 129-1239 129-1240 129-1241 129-1243 129-1245 129-1247 129-1249 129-1250 129-1251 129-1252 129-1255 129-1259 129-1261
Sterlisation flat pouches, Stericlin®
Beutel Sterilisationsbeutel
These see-through sterilisation flat pouches made of sterilisation proof 70 g/m² heavy-duty paper are a universal packaging solution.
  • Effective barrier against germs and bacteria
  • Permits the transmission of air, steam or sterilisation gases
  • Three indicators for STEAM, EO and FORM sterilisation are imprinted outside the filling space
Suitable for sterilisation applications of almost all light and medium weight instruments and sets.
Standards: DIN EN ISO 11607 and DIN 868-5.