Sustainable 2-in-1 flat mop covers, Revolve™
Lieferant: Texwipe
TX1720 TX1721
129-1777EA 1340 EUR
129-1777 129-1778
Sustainable 2-in-1 flat mop covers, Revolve™
Mopps
Revolve™ 2-in-1 mop covers, the first sustainable covers made for the cleanroom industry, are made from 100% upcycled polyester. Revolve™ mop covers contribute to sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing the level of carbon emissions.
  • Mop and clean large and small surfaces
  • Apply and remove disinfectants
  • Spill control
  • 100% upcycled polyester
  • Clean with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
  • Appropriate for use with temperatures less than 400 °F (205 °C)
The integrated polyester cover/pad design allows replacing covers and pads in a one-step process. The integrated design offers high sorption capacity, which is ideal for spill control, cleaning, and solution application. The covers easily fit over the mop head, permitting a quick change. The integrated polyester pad assures the mop cover conforms to surfaces for consistent contact and cleaning. Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles, and fibres for use in critical cleaning applications and environments. Autoclave safe. Individually lot coded for traceability and quality control.
Standards: Non-sterile wipes are compatible for use in ISO class 3 to 7 / GMP grade B to D.