High-Capacity Cleaning Cart
Lieferant: Rubbermaid Commercial Products
FG9T7200BLA
129-1948EA 1060 EUR
129-1948
High-Capacity Cleaning Cart
Transportwagen
The Rubbermaid Commercial housekeeping high-capacity cleaning cart is a customisable solution with room for additional add-ons and accessories to meet your needs.
  • Increased efficiency
  • Reduce footprint easily
  • Customisable storage space
  • Easy access to stick goods and safety signs
  • Enhance facility image
  • Seamless maneuverability
  • Durable commercial-grade construction; sleek and functional design
The trolley offers 40% more space than traditional cleaning trolleys to minimise trips for additional supplies and maximises staff productivity. Interchangeable handles and removable platform allow user to easily expand or reduce the footprint of the trolley to meet specific cleaning and storage needs. Expansive storage space with removable bins and caddies let staff restock before they clean, increasing productivity by eliminating time spent searching for additional supplies. Organise and stock cleaning supplies in a way that best suits your needs. Rubber tool grips and hooks securely hold up to five stick goods, while hooks hold up to four safety signs.
The durable vinyl bag hides rubbish from patrons, making it great for cleaning high-profile areas. The smooth surface and detachable vinyl zipped bag makes it easy to clean, which helps keep it looking new. Durable 200 mm ball-bearing wheels and quiet 100 mm casters make for quiet, easy mobility for workers. Water- and chemical-resistant finish provides easy maintenance and long-lasting good looks. Mop bucket platform with securing grooves keeps WaveBrake or microfibre charging buckets securely attached to trolley.
Lieferumfang: Includes 2 removable 9 L disinfecting caddies, 2 swivel casters and 2 wheels and 1 high capacity yellow vinyl bag, 128 L.