Doypack® Stand-up Paper/Aluminium/PE Zip Seal Bags
Lieferant: Flexico
ITM080303 ITM080302 ITM080304 ITM080305
MINIITM080303EA 408 EUR
MINIITM080303 129-3898 129-3899 129-3900
Doypack® Stand-up Paper/Aluminium/PE Zip Seal Bags
Beutel Aufbewahrungsbeutel
Zip seal bags designed for food contact.
  • Moisture and oxygen barrier
  • Ideal for dry, humid, and aqueous products
  • Suitable for food contact
  • Zip seal closure with sound and tactile feel
  • Resealable and reusable
  • Upright position for shelf display
  • Bottom gusset : 2×35 mm
The multi-layer paper/aluminium/polyethylene film provides excellent preservation for bag contents. The kraft paper gives the bag an authentic and natural look. The aluminium layer ensures preservation of product freshness and gives it the best protection against humidity, UV, and oxygen.More Product Information