Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12909.09
12909.18
12909.30
12909.A1
12909.09EA
36.5
EUR
12909.09
12909.18
12909.30
12909.A1
Praseodymtrinitrat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Praseodymtrinitrat Hydrat
Praseodymium(III) nitrate hydrate is used in fluorescent display tubes and phosphors. It is also used in the ultrasonic synthesis of phase-pure praseodymium molybdate. It plays a major role in the solvothermal process for formation of doped lanthanide oxysulfide
Purity: 99.9%
CAS No.: 15878-77-0
Molecular Formula: Pr(NO3)3.xH2O
Formula Weight: 326.92(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Pale green
Odour: Odourless
UN No.: 1477
MDL No.: MFCD00011175
Formel:
N₃O₉Pr Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011175 CAS-Nummer: 15878-77-0 EINECS: 233-796-5 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...