Nickel ≥99,999% (Metall-Basis), Pulver, Puratronic® (max. 100ppm C)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
12966.14
12966.22
12966.A1
12966.14EA
154
EUR
12966.14
12966.22
12966.A1
Nickel ≥99,999% (Metall-Basis), Pulver, Puratronic® (max. 100ppm C)
Nickel
Purity: 99.999%
CAS No.: 7440-02-0
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00011137
Formel:
Ni Siedepunkt: 2371 °C (1013 hPa) Schmelzpunkt: 1455 °C Dichte: 8,9 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011137 CAS-Nummer: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...