Lithium acetate hydrate ≥99,998% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific

12982.06 12982.14
12982.06EA 47 EUR
12982.06 12982.14
Lithium acetate hydrate ≥99,998% (Metall-Basis), Puratronic®
Lithium acetate hydrate

Purity: 99.998% CAS No.: 546-89-4 Molecular Formula: LiOOCCH3.xH2O Formula Weight: 65.98(anhy) Physical Form: Crystalline Melting Point: 57.8° MDL No.: MFCD00013057

Formel: H₃CCOOLi·xH₂O
Schmelzpunkt: 291 °C
Dichte: 1,26 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013057
CAS-Nummer: 546-89-4
EINECS: 208-914-3
Merck Index: 14,5521

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR