Amiloride hydrochloride, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Amiloride hydrochloride hydrate, 3,5-Diamino-N-carbamimidoyl-6-chloropyrazine-2-carboxamide hydrochloride, Midamor, Colectril, Amipramizide, Guanamprazine hydrochloride, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, 1-(3,5-diamino-2-chloropyrazine-6-carbonyl)guanidine hydrochloride
This is an inhibitor of angiogenesis. Inhibits capillary morphogenesis completely and reversibly at approximately 130 µM. At low concentrations, acts as a potent and specific inhibitor of transmembrane Na+ entry and Na+, K+-ATPase. Blocks smooth muscle responses to contractile stimuli and palytoxin-induced conductance in skeletal muscle.
- Acts as an inhibitor of urokinase-type plasminogen activator (uPA)
- At higher concentrations, blocks the Na+/H+ exchange pathway
- Reduces electrical potential across tubular epithelium
Formel:
C₆H₈ClN₇O·HCl Molecular Weight: 266,09 g/mol |
MDL:
MFCD00058197 CAS-Nummer: 2016-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...