Calciumwolframat ≥98%

Lieferant: Thermo Fisher Scientific

13051.14 13051.22 13051.36
13051.14EA 64 EUR
13051.14 13051.22 13051.36
Calciumwolframat ≥98%
Calciumwolframat

Purity: 98% CAS No.: 7790-75-2 Molecular Formula: CaWO4 Formula Weight: 287.93 Physical Form: -325 Mesh Powder Melting Point: 1620° MDL No.: MFCD00010913

Formel: CaO₄W
Schmelzpunkt: 1620 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010913
CAS-Nummer: 7790-75-2
EINECS: 232-219-4
Merck Index: 14,1712

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR