Blei(IV)fluorid 99
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bleitetrafluorid
13070.04
13070.04EA
110
EUR
13070.04
Blei(IV)fluorid 99
Blei(IV)fluorid
Purity: 99%
CAS No.: 7783-59-7
Molecular Formula: PbF4
Formula Weight: 283.18
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
UN No.: 2291
MDL No.: MFCD00016278
Formel:
F₄Pb Molecular Weight: 283.19 g/mol Schmelzpunkt: 600 Dichte: 6.7 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016278 CAS-Nummer: 7783-59-7 EINECS: 232-012-9 UN: 2291 Merck Index: 12,05448 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...