Titan(III)fluorid

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Titantrifluorid

13079.09 13079.18
13079.09EA 75.7 EUR
13079.09 13079.18
Titan(III)fluorid
Titan(III)fluorid

CAS No.: 13470-08-1 Molecular Formula: TiF3 Formula Weight: 104.90 Physical Form: Powder Density: 3.400 Sensitivity: Air and Moisture Sensitive UN No.: 3260 MDL No.: MFCD00042544

Formel: F₃Ti
Dichte: 3,400
Lagertemperatur: Raumtemperatur
MDL: MFCD00042544
CAS-Nummer: 13470-08-1
EINECS: 236-732-4
UN: 3260
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR