Titan(III)fluorid
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Titantrifluorid
13079.09
13079.18
13079.09EA
75.7
EUR
13079.09
13079.18
Titan(III)fluorid
Titan(III)fluorid
CAS No.: 13470-08-1
Molecular Formula: TiF3
Formula Weight: 104.90
Physical Form: Powder
Density: 3.400
Sensitivity: Air and Moisture Sensitive
UN No.: 3260
MDL No.: MFCD00042544
Formel:
F₃Ti Dichte: 3,400 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042544 CAS-Nummer: 13470-08-1 EINECS: 236-732-4 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...