4-Phenylazophenol ≥98%, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
|
M-Clarity™ Qualitätsstufe = MQ100
4-Phenylazophenol is a polymethacrylates containing polar azo dye. 4-Phenylazophenol (4PP) has been used to study mineralisation by P. chrysosporium under nitrogen-sufficient and nitrogen-limiting culture conditions.
Formel:
C₁₂H₁₀N₂O Schmelzpunkt: 150,..152 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00002330 CAS-Nummer: 1689-82-3 EINECS: 216-880-6 |
|
Spezifikation Testergebnisse
Appearance (Color) | Yellow to Gold |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Wavelength | 233 - 239 nm |
Extinction Coefficient | ≥22000 |
Extinction Coefficient, c = 0.005g/l, Methanol | ≥10000 |
Carbon | 70.9 - 74.5 % |
Nitrogen | 13.7 - 14.5 % |
Purity (TLC) | ≥98 % |
Solubility (Turbidity), c = 0.1%, Ethanol | clear |
Solubility (Color), Dark Yellow to Orange-Brown | Conforms to Requirements |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...