Aluminiumselenid ≥99% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Aluminum selenide
13132.03
13132.03EA
90.8
EUR
13132.03
Aluminiumselenid ≥99% (Metall-Basis)
Aluminium selenide
Purity: 99%
CAS No.: 1302-82-5
Molecular Formula: Al2Se3
Formula Weight: 290.84
Physical Form: 6.5mm & Down Pieces
Density: 3.437
Appearance: Pale brown
Odour: Odourless
Solubility: Decomposes to Al203 and SeO2 in water
Merck Reference: 14,361
UN No.: 3283
MDL No.: MFCD00014158
Formel:
Al₂Se₃ Molecular Weight: 290,84 g/mol Schmelzpunkt: 960 °C Dichte: 3,437 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014158 CAS-Nummer: 1302-82-5 EINECS: 215-110-6 UN: 3283 ADR: 6.1,II Merck Index: 14,361 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...