Neodym(III)sulfat Octahydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Scientific
|
Achtung
|
13185.14
13185.14EA
298
EUR
13185.14
Neodym(III)sulfat Octahydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Neodym(III)sulfat Octahydrat
Purity: 99.99%
CAS No.: 13477-91-3
Molecular Formula: Nd2(SO4)3.8H2O
Formula Weight: 720.78 (576.66anhy)
Physical Form: Crystalline
Density: 2.85
Melting Point: 1176°
Merck Reference: 14,6450
MDL No.: MFCD00149803
Formel:
Nd₂S₃O₁₂·8H₂O Molecular Weight: 720,78 g/mol (576,66 anhy) Schmelzpunkt: 1176 °C Dichte: 2,85 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149803 CAS-Nummer: 13477-91-3 EINECS: 233-262-1 Merck Index: 14,6450 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...