Lanthanum(III) 2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13194.14
13194.22
13194.14EA
118
EUR
13194.14
13194.22
Lanthanum(III) 2,4-pentanedionate hydrate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lanthanum(III) 2,4-pentanedionate hydrate
Purity: 99.9%
CAS No.: 64424-12-0
Molecular Formula: C15H21LaO6.xH2O
Formula Weight: 436.24(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale yellow
Melting Point: 143°
MDL No.: MFCD00149057
Formel:
C₁₅H₂₁LaO₆·xH₂O Schmelzpunkt: 143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149057 CAS-Nummer: 64424-12-0 EINECS: 238-187-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...