Lanthanum(III) oxalate decahydrate ≥99,99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13196.18
13196.18EA
81.6
EUR
13196.18
Lanthanum(III) oxalate decahydrate ≥99,99%
Lanthanum(III) oxalate decahydrate
Purity: 99.99%
CAS No.: 537-03-1
Molecular Formula: C6La2O12.10H2O
Formula Weight: 722.02 (541.87anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point: dec.
UN No.: UN3288
MDL No.: MFCD00150540
Formel:
C₆La₂O₁₂·10H₂O Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150540 CAS-Nummer: 537-03-1 EINECS: 208-656-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...