Redox standards, Supelco®
Lieferant: Merck
78855-500ML
80138-500ML
87783-500ML
13198-500ML
04887-500ML
40661-500ML
78855-500MLEA
126
EUR
78855-500ML
80138-500ML
87783-500ML
13198-500ML
04887-500ML
40661-500ML
Redox standards, Supelco®
Standards
Redox-/ORP-Standards
The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.