Gadolinium(III)bromid Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®

Lieferant: Thermo Scientific
Achtung

13208.22
13208.22EA 583 EUR
13208.22
Gadolinium(III)bromid Hydrat ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Gadolinium(III)bromid Hydrat

Purity: 99.99% CAS No.: 30010-20-9 Molecular Formula: GdBr3.xH2O Formula Weight: 396.96(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00049741

Formel: Br₃Gd·H₂O
Molecular Weight: 414,98 g/mol
Schmelzpunkt: 186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049741
CAS-Nummer: 30010-20-9
EINECS: 237-494-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR