Lithiumsulfat, wasserfrei ≥99,7% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

13404.30 13404.A1 13404.A7
13404.30EA 71 EUR
13404.30 13404.A1 13404.A7
Lithiumsulfat, wasserfrei ≥99,7% (Metall-Basis)
Lithium sulphate

Purity: 99.7% CAS No.: 10377-48-7 Molecular Formula: Li2SO4 Formula Weight: 109.94 Physical Form: -40 Mesh Powder Density: 2.22 Appearance: White Odour: Odourless Melting Point: 845° Merck Reference: 14,5541 MDL No.: MFCD00011086

Formel: Li₂O₄S
Siedepunkt: 1377 °C (1013 hPa)
Schmelzpunkt: 845 °C
Dichte: 2,22
Lagertemperatur: Raumtemperatur
MDL: MFCD00011086
CAS-Nummer: 10377-48-7
EINECS: 233-820-4
Merck Index: 14,5541

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR