Natriumcarbonat, anhydrous 99,5-100,5 Dried, Puriss. NF, Ph. Eur., BP, FCC, meetSpec_BP,Ph.Eur.,NF,FCC,E500, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: wasserfreie Soda

13418-5KG 13418-2.5KG 13418-25KG 13418.9025 13418-1KG
13418-5KGEA 249.13 EUR
13418-5KG 13418-2.5KG 13418-25KG HONE13418.9025 13418-1KG
Natriumcarbonat, anhydrous 99,5-100,5 Dried, Puriss. NF, Ph. Eur., BP, FCC, meetSpec_BP,Ph.Eur.,NF,FCC,E500, Fluka™
Natriumcarbonat
Formel: Na₂CO₃
Molecular Weight: 105.99 g/mol
Siedepunkt: 1600
Schmelzpunkt: 854 °C
Dichte: 2.53 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003494
CAS-Nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated to the dried substance) 99.5 - 100.5 %
Loss on drying (300°C) Max. 0.5 %
Loss on drying (105°C, 4 h) Max. 0.5 %
Arsenic (As) Max. 2 ppm
Copper (Cu) Max. 0.002 %
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.002 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 50 ppm
Alkali hydroxide, -bicarbonates complying
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR