Lithiumcarbonat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13418.36
13418.A3
13418.A9
13418.36EA
158
EUR
13418.36
13418.A3
13418.A9
Lithiumcarbonat ≥99%
Lithium carbonate
Purity: 99%
CAS No.: 554-13-2
Molecular Formula: Li2CO3
Formula Weight: 73.90
Physical Form: Powder
Density: 2.11
Appearance: White
Odour: Odourless
pH: 10-11.5
Melting Point: 723°
Boiling Point: 1310°dec.
Solubility: Soluble in water and dilute acids. Insoluble in alcohol
Merck Reference: 14,5527
MDL No.: MFCD00011084
Formel:
Li₂CO₃ Molecular Weight: 73.89 g/mol Siedepunkt: 1310 °C (Zers.) Schmelzpunkt: 723 °C Dichte: 2.11 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011084 CAS-Nummer: 554-13-2 EINECS: 209-062-5 Merck Index: 14,5527 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...