di-Lithiumtetraborat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13419.18
13419.30
13419.A1
13419.18EA
41.1
EUR
13419.18
13419.30
13419.A1
di-Lithiumtetraborat ≥99% (Metall-Basis)
di-Lithiumtetraborat
Purity: 99%
CAS No.: 12007-60-2
Molecular Formula: B4Li2O7
Formula Weight: 169.12
Physical Form: Powder
Density: 2.4
Appearance: White
Odour: Odourless
Melting Point: 930°
Merck Reference: 14,5524
MDL No.: MFCD00011083
Formel:
Li₂B₄O₇ Schmelzpunkt: 930 °C Dichte: 2,4 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011083 CAS-Nummer: 12007-60-2 EINECS: 234-514-3 Merck Index: 14,5524 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...