Lithiumperoxid ≥95%

Lieferant: Thermo Fisher Scientific
Gefahr

13421.22 13421.36
13421.22EA 448 EUR
13421.22 13421.36
Lithiumperoxid ≥95%
Lithiumperoxid

Purity: 95% CAS No.: 12031-80-0 Molecular Formula: Li2O2 Formula Weight: 45.88 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White to off-white Odour: Odourless UN No.: 1472 MDL No.: MFCD00016184

Formel: Li₂O₂
Dichte: 1,2063 g/cm³ (18 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016184
CAS-Nummer: 12031-80-0
EINECS: 234-758-0
UN: 1472
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR