Natriumhydrid 57 - 63% Dispersion in Mineralöl
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
13431.18
13431.36
13431.18EA
43.5
EUR
13431.18
13431.36
Natriumhydrid 57 - 63% Dispersion in Mineralöl
Sodium hydride
CAS No.: 7646-69-7
Molecular Formula: HNa
Formula Weight: 24.00
Physical Form: Moist powder
Density: 0.92
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white to gray
Odour: Odourless
Merck Reference: 14,8625
UN No.: 1427
MDL No.: MFCD00003471
Formel:
HNa Dichte: 0,92 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003471 CAS-Nummer: 7646-69-7 EINECS: 231-587-3 UN: 1427 ADR: 4.3,I Merck Index: 14,8625 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...