Natriumborhydrid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
13432.22
13432.36
13432.57
13432.22EA
82.9
EUR
13432.22
13432.36
13432.57
Natriumborhydrid ≥98%
Natriumtetrahydroborat
Purity: 98%
CAS No.: 16940-66-2
Molecular Formula: NaBH4
Formula Weight: 37.83
Physical Form: Powder
Density: 1.074
Appearance: White
Odour: Weak
Melting Point: 400° dec.
Merck Reference: 14,8592
UN No.: 1426
MDL No.: MFCD00003518
Formel:
NaBH₄ Schmelzpunkt: 400 °C (Zersetzung) Dichte: 1,074 Flammpunkt: 69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003518 CAS-Nummer: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3,I Merck Index: 14,8592 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...