Lithiumaluminiumhydrid ≥95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 95% min
CAS No.: 16853-85-3
Molecular Formula: LiAlH4
Formula Weight: 37.95
Physical Form: Powder
Density: 0.917
Appearance: White to grey
Odour: Odourless
Melting Point:
Reducing agent for pharmaceuticals, perfumes, and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizations
Formel:
H₄AlLi Schmelzpunkt: >125 °C (Zersetzung) Dichte: 0,917 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011075 CAS-Nummer: 16853-85-3 EINECS: 240-877-9 UN: 1410 ADR: 4.3,I Merck Index: 14,349 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...