Kaliumnitrat ≥99,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Kalisalpeter
13443.22
13443.36
13443.A3
13443.22EA
27.2
EUR
13443.22
13443.36
13443.A3
Kaliumnitrat ≥99,0% ACS
Kaliumnitrat
Purity: 99.0% min
CAS No.: 7757-79-1
Molecular Formula: KNO3
Formula Weight: 101.11
Physical Form: Crystalline
Density: 2.109
Appearance: White
Odour: Odourless
Melting Point: 334°
Boiling Point: 400° dec.
Solubility: Soluble in water, glycerol, liquid NH3
Merck Reference: 14,7648
UN No.: 1486
MDL No.: MFCD00011409
Pyrotechnics, glass manufacture, tempering steel, reagent
Formel:
KNO₃ Siedepunkt: 400 °C (Zersetzung) Schmelzpunkt: 334 °C Dichte: 2,109 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011409 CAS-Nummer: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 14,7648 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...