Kaliumdichromat ≥99,0% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

13450.22 13450.36 13450.A4
13450.22EA 40.5 EUR
13450.22 13450.36 13450.A4
Kaliumdichromat ≥99,0% ACS
Kaliumdichromat

Purity: 99.0% min CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Crystalline Density: 2.676 Appearance: Orange Odour: Odourless Melting Point: 398° Solubility: Soluble in water Merck Reference: 14,7627 UN No.: 3087 MDL No.: MFCD00011367


As an oxidizing agent, in dyeing, painting, decorating porcelain, lithography, in electric batteries, electroplating

Formel: K₂Cr₂O₇
Siedepunkt: 500 °C (1013 hPa)
Schmelzpunkt: 398 °C
Dichte: 2,676
Lagertemperatur: Raumtemperatur
MDL: MFCD00011367
CAS-Nummer: 7778-50-9
EINECS: 231-906-6
UN: 3087
ADR: 5.1,II
REACH: 01-2119454792-32
Merck Index: 14,7627

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR