Rubidium formate hydrate ≥99,8% (Metall-Basis)
Lieferant: Thermo Scientific
|
Achtung
|
13499.09
13499.09EA
59.6
EUR
13499.09
Rubidium formate hydrate ≥99,8% (Metall-Basis)
Rubidium formate hydrate
Purity: 99.8%
CAS No.: 3495-35-0
Molecular Formula: CHO2Rb.xH2O
Formula Weight: 130.49(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00054359
Formel:
CHO₂Rb·xH₂O Molecular Weight: 130,49 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054359 CAS-Nummer: 3495-35-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...