Pharmaceutical exhausts
Lieferant: asecos
31833 31834
135-1772EA 2637 EUR
135-1772 135-1773
Pharmaceutical exhausts
Dunstabzugshauben und Verkleidungen Apotheken-Abzüge
For safe handling of chemicals, protection from hazardous vapours and potentially explosive atmospheres. Hazardous material workplace (pharmaceutical exhausts).
  • Easily mounted on a table top
  • Robust construction made of chemically resistant, antibacterial plastic laminated sheet steel
  • Exhaust air monitoring with visual and audible alarm
Standard equipment: exhaust air monitoring, 2 sockets, workplace lighting, acrylic glass sliding windows with continuously adjustable height, exhaust air fan. Accessories like media supplies, support frame and ventilation sets are available on request.
Standard equipment: exhaust air monitoring, 2 sockets, workplace lighting, acrylic glass sliding windows with coninuously adjustable height, exhaust air fan. Accessories like media supplies, support frame and ventilation sets are available on request.
Pressure drop with extraction air duct: 315 PA
Standards: Conform to DIN 12924 Part 4: 2012 CE. Made in Germany. Testing of the air equipment by recognised test institute
Lieferumfang: Delivery prices avaible upon request.