Cabinets for chemicals, CS-CLASSIC
Lieferant: asecos
30803050
135-1938EA 1939 EUR
135-1938
Cabinets for chemicals, CS-CLASSIC
Möbel Lagerschränke
For safe, approved and separate storage of different hazardous materials in one cabinet.
  • Door variants: wing doors with integrated ventilation system for each compartment
  • Natural ventilation at the bottom (vent openings at the cabinet bottom)
  • Connection (NW 75) to a technical exhaust system on the top
  • Double-walled door construction
  • High-quality, scratch-resistant structured paint finish
  • Safety cylinder lock integrated in the turning handle
  • Adjusting aids to compensate for uneven floor
Made in Germany.
Outer body made sturdy all welded construction suitable for highest industrial demands with the attached labelling system.
Bestellinformation: Door colours are available on request. Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition. Please check the website or contact Avantor sales office for further information.
Lieferumfang: Delivery prices available upon request.