Safety storage cabinets, under bench, Type UB-T-30, UB-line
Lieferant: asecos
30445001
135-2075EA 1767 EUR
135-2075
Safety storage cabinets, under bench, Type UB-T-30, UB-line
Möbel Lagerschränke
Under bench cabinets with wing doors. Fire resistance: 30 minutes.
  • Safety access to all containers, interior of drawers are completely visible
  • Drawers are lockable with cylinder lock
  • Flexible and mobile – castors with plinth optionally available
  • Integrated air ducts ready for connection (NW 75) to a technical exhaust system technical
  • Convenient handling doors which can be opened with minimal effort
Standards: EN 14727 conformity (laboratory furniture), GS approved, CE conformity. Furnace tested (type test) in accordance with EN 14470-1
Bestellinformation: Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition.
Lieferumfang: Delivery prices available upon request.