Safe passive storage of lithium-ion batteries.
- Robust construction and longevity; Convenient cabinet entry
- Integrated potential-free alarm contact; No unauthorised use
- Integrated transport base; Easy alignment
- Smoke detector and temperature sensor; Pressure relief
- All-around protection
- LED display
- Fire suppression system
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface.
Convenient cabinet entry: open both doors at the same time with just one hand. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with relay module and module for remote signalling.
No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green). Permanent locking of the doors by automatic 2-point locking. Integrated transport base: In the event of an emergency, immediate evacuation and transport of the cabinet to a safe place outdoors.
Easy alignment: adjusting aids to compensate for uneven floor. Smoke detector and temperature sensor for fire detection and forwarding the alarm to the building management system. Pressure relief in the head section.
All-around protection: 90 minutes fire protection from outside to inside (type 90/type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside. LED display with visual and audible alarm for a quick indication of emergencies without opening the cabinet doors. Fire suppression system: Triggers in the event of a fire inside the cabinet automatically.
Standards: Fire resistance 90 minutes. Type tested in accordance with EN 14470-1. CE compliant.
Zubehörinformationen: The optional relay module (sold separately) can differentiate between 4 different warning and alarm levels (Warning, Alarm level 1, Alarm level 2 and power failure) and passes them on to a control centre via separate potential-free contacts either as make contacts or break contacts 1× connector for customer wiring for one of the 4 warning and alarm levels is included in the delivery.
Note: The switching contacts are used to connect a signal to a building management system. Direct integration into a fire alarm control panel (FACP) is not recommended or may only be carried out in consultation with the person responsible for the system.
Bestellinformation: Interior equipment packages need to be ordered separately (not included). Please contact your Avantor representative for more information and available options.
Lieferumfang: Delivery prices available upon request.
Bitte beachten According to the security concept it is highly recommended to place ION-LINE cabinets at the ground level in a location from where a quick evacuation from the building is possible in the event of fire.