Praseodymtrifluorid, anhydrous 99,9 REO basis, REacton®

Lieferant: Thermo Fisher Scientific
Gefahr

13658.14
13658.14EA 133 EUR
13658.14
Praseodymtrifluorid, anhydrous 99,9 REO basis, REacton®
Praseodymtrifluorid

Purity: 99.9% CAS No.: 13709-46-1 Molecular Formula: PrF3 Formula Weight: 197.90 Physical Form: Crystalline Density: 6.2 Appearance: Green Odour: Odourless Melting Point: 1395° UN No.: 3288 MDL No.: MFCD00016287

Formel: F₃Pr
Molecular Weight: 197.9 g/mol
Schmelzpunkt: 1395
Dichte: 6.2
Lagertemperatur: Raumtemperatur
MDL: MFCD00016287
CAS-Nummer: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR