Cobalt(II)chlorid, ultra trocken ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

13666.03 13666.06 13666.14
13666.03EA 69.1 EUR
13666.03 13666.06 13666.14
Cobalt(II)chlorid, ultra trocken ≥99,9% (Metall-Basis)
Cobalt(II)chlorid

Purity: 99.9% CAS No.: 7646-79-9 Molecular Formula: Cl2Co Formula Weight: 129.84 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.367 Appearance: Pale blue Odour: Odourless Melting Point: 735° Boiling Point: 1049° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00010938

Formel: Cl₂Co
Siedepunkt: 1049 °C
Schmelzpunkt: 735 °C
Dichte: 3,367
Lagertemperatur: Raumtemperatur
MDL: MFCD00010938
CAS-Nummer: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR