trans-Bis(triphenylphosphin)palladium(II)bromid (13,4% Pd)
Lieferant: Thermo Fisher Scientific
CAS No.: 22180-53-6
Molecular Formula: Br2Pd[P(C6H5)3]2
Formula Weight: 790.80
Physical Form: Powder
Sensitivity: Hygroscopic
Solubility: Soluble in chloroform, toluene and benzene
MDL No.: MFCD00134175
Hydrogenation
Formel:
C₃₆H₃₂Br₂P₂Pd Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134175 CAS-Nummer: 22180-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...