Chloropentaammincobalt(III)chlorid
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Slightly soluble in water, soluble in conc. H₂SO₄. Insoluble in alcohol
CAS No.: 13859-51-3
Molecular Formula: H15Cl3CoN5
Formula Weight: 250.44
Physical Form: Powder
Density: 1.819
Appearance: Purple
Odour: Odourless
Melting Point: dec.
Solubility: Slightly soluble in water, soluble in conc. H2SO4. Insoluble in alcohol
MDL No.: MFCD00064728
Formel:
H₁₅Cl₃CoN₅ Dichte: 1,819 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064728 CAS-Nummer: 13859-51-3 EINECS: 237-594-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...