Cadmium-Plasma-Standardlösungen für ICP, Specpure®
Lieferant: Thermo Fisher Scientific
13813.AD
13813.AE
13813.AP
14424.AE
14424.AP
13813.ADEA
53.7
EUR
13813.AD
13813.AE
13813.AP
14424.AE
14424.AP
Cadmium-Plasma-Standardlösungen für ICP, Specpure®
Standards
Kationischer Standards
Cadmium-Standards
Molecular Formula: Cd in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00010914
Standards: Rückführbar auf NIST
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...