Blei, Plasma-Standardlösungen, Specpure®
Lieferant: Thermo Fisher Scientific
13853.AD
13853.AE
13853.AP
14412.AD
14412.AE
14412.AP
13853.ADEA
54.4
EUR
13853.AD
13853.AE
13853.AP
14412.AD
14412.AE
14412.AP
Blei, Plasma-Standardlösungen, Specpure®
Standards
Kationischer Standards
Blei-Standards
Pb in 5% HNO₃
UN Nr.: 3264
MDL Nr.: MFCD00134050
Standards: Rückführbar auf NIST
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...