Reinecke-Salz, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor

1388.0025EA 393.69 EUR
1388.0025
Reinecke-Salz, BAKER ANALYZED® ACS, J.T.Baker®
Reinecke-Salz
Formel: C₄H₁₀CrN₇S₄
MDL: MFCD00151654
CAS-Nummer: 13573-16-5
EINECS: 237-003-3

Bestellen

Spezifikation Testergebnisse

Meets ACS Specifications
Assay min. 93.0 %
Insoluble in Dilute HCl max. 0.05 %
Sensitivity passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR