Sigma-Sil-A, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

1391-10X1ML 1391-1ML 1391-5X1ML
1391-10X1MLEA 544 EUR
1391-10X1ML 1391-1ML 1391-5X1ML
Sigma-Sil-A, Sigma-Aldrich®
Sigma-Sil-A

Trimethylchlorosilane: hexamethyldisilazane: pyridine (1:3:9) reagent for preparing TMS derivatives of amino acids, sugars, etc.

Lagertemperatur: Raumtemperatur
MDL: MFCD00080961
ADR: 3 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR