EDTA Dinatriumsalz Dihydrat 99,0-101,0% USP, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Avantor excipients are used during the formulation process in a wide range of applications. Avantor provides high-purity and performance excipients that serve as fillers, binders, disintegrants, lubicants, flavors, emulsifiers and preservatives. All the products are cGMP produced and meet USP grade standards.
Formel:
C₁₀H₁₄N₂Na₂O₈·2H₂O |
MDL:
MFCD00150037 CAS-Nummer: 6381-92-6 |
|
Spezifikation Testergebnisse
Meets U.S.P Requirements | — |
GMP Manufactured Product | — |
CAUTION: For Manufacturing, processing or repackaging | — |
Bulk Pharmaceutical Chemical | — |
USP - Assay (C₁₀H₁₄N₂Na₂O₈) (anhydrous basis) | 99.0 - 101.0 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Identification C | Passes Test |
USP - pH (1 in 20) | 4.0 - 6.0 |
USP - Loss on Drying | 8.7 - 11.4 % |
USP - Calcium (Ca) | Passes Test |
USP - Limit of Nitrilotriacetic Acid | ≤0.1 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Meets Reagent Specifications for testing USP/NF monographs | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...