Cyclopentadienylthallium

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Thallium(I) cyclopentadienide

13958.06 13958.14
13958.06EA 83 EUR
13958.06 13958.14
Cyclopentadienylthallium
Cyclopentadienylthallium

CAS No.: 34822-90-7 Molecular Formula: C5H5Tl Formula Weight: 269.47 Physical Form: Powder Sensitivity: Air Sensitive Melting Point: dec. UN No.: 1707 MDL No.: MFCD00134141

Formel: C₅H₅Tl
Molecular Weight: 269.48 g/mol
Siedepunkt: 90…110 °C (1 mmHg)
Schmelzpunkt: 75 Zersetzung
Lagertemperatur: Raumtemperatur
MDL: MFCD00134141
CAS-Nummer: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR