Praseodym ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 1.0 mm (0.04 in)

Lieferant: Thermo Fisher Scientific

13963.FF 13963.FI
13963.FFEA 211 EUR
13963.FF 13963.FI
Praseodym ≥99,5% (REO, Basis der Oxide der Metalle der seltenen Erden), Folie, Dicke 1.0 mm (0.04 in)
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Physical Form: ≈4.23g/25x25mm, Packaged in mylar under argon Appearance: White Odour: Odourless MDL No.: MFCD00011174

Formel: Pr
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR