Titan ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13976.FF
13976.FI
13976.GH
13976.FFEA
65.9
EUR
13976.FF
13976.FI
13976.GH
Titan ≥99,99% (Metall-Basis), Folie, Dicke 0.127 mm (0.005 in)
Titanium
Purity: 99.99+%
CAS No.: 7440-32-6
Physical Form: ≈0.36g/25x25mm
Odour: Odourless
MDL No.: MFCD00011264
Formel:
Ti Molecular Weight: 47.88 g/mol Siedepunkt: 3262 Schmelzpunkt: 1660 Dichte: 4.51 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011264 CAS-Nummer: 7440-32-6 EINECS: 231-142-3 Merck Index: 12,09610 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...