Gallium(III)fluorid ≥99,995% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
13987.03
13987.06
13987.03EA
168
EUR
13987.03
13987.06
Gallium(III)fluorid ≥99,995% (Metall-Basis)
Gallium trifluoride
Purity: 99.999%
CAS No.: 7783-51-9
Molecular Formula: GaF3
Formula Weight: 126.72
Physical Form: -20 Mesh Powder
Density: 4.47
Appearance: White
Odour: Odourless
Melting Point: 800° subl.
Boiling Point: ca 1000°
Merck Reference: 14,4354
UN No.: 3260
MDL No.: MFCD00011021
Formel:
F₃Ga Siedepunkt: ∼1000 °C Schmelzpunkt: 800 °C (Subl.) Dichte: 4,47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011021 CAS-Nummer: 7783-51-9 EINECS: 232-004-5 UN: 3260 ADR: 8,III Merck Index: 14,4354 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...