Cobalt(IV)sulfid ≥99,5% (Metall-Basis ausgeschlossen Ni) (max. 0,2% Ni)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cobaltdisulfid
14005.06
14005.06EA
135
EUR
14005.06
Cobalt(IV)sulfid ≥99,5% (Metall-Basis ausgeschlossen Ni) (max. 0,2% Ni)
Cobalt disulphide
Purity: 99.5%
CAS No.: 12013-10-4
Molecular Formula: CoS2 (may contain traces of other Co-S phases)
Formula Weight: 123.06
Physical Form: -200 Mesh Powder
Appearance: Black
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00064735
Formel:
CoS₂ Molecular Weight: 123,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064735 CAS-Nummer: 12013-10-4 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...