Germanium(IV)iodid ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
14009.09
14009.18
14009.09EA
333
EUR
14009.09
14009.18
Germanium(IV)iodid ≥99,999% (Metall-Basis)
Germanium(IV)iodid
Purity: 99.999%
CAS No.: 13450-95-8
Molecular Formula: GeI4
Formula Weight: 580.21
Physical Form: -10 Mesh Crystalline
Density: 4.32
Melting Point: 146°
UN No.: 3260
MDL No.: MFCD00016120
Formel:
GeI₄ Siedepunkt: 350 °C (1013 hPa) Schmelzpunkt: 146 °C Dichte: 4,32 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016120 CAS-Nummer: 13450-95-8 EINECS: 236-613-7 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...