Tantalpentafluorid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

14052.09 14052.18
14052.09EA 187 EUR
14052.09 14052.18
Tantalpentafluorid ≥99,9% (Metall-Basis)
Tantalpentafluorid

Purity: 99.9% CAS No.: 7783-71-3 Molecular Formula: TaF5 Formula Weight: 275.94 Physical Form: Powder Density: 4.74 Appearance: White Melting Point: 96.8° Boiling Point: 229.5° Solubility: Decomposes in water forming tantalum oxyfluoride. Soluble in conc. and fuming HNO3 Merck Reference: 14,9055 UN No.: 3260 MDL No.: MFCD00042542

Formel: F₅Ta
Siedepunkt: 229,5 °C
Schmelzpunkt: 96,8 °C
Dichte: 4,74
Lagertemperatur: Raumtemperatur
MDL: MFCD00042542
CAS-Nummer: 7783-71-3
EINECS: 232-022-3
UN: 3260
ADR: 8,II
Merck Index: 14,9055

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR